[(2-Oxo-2-piperidin-1-ylethyl)(phenyl)amino]acetic acid

Supplier: Matrix Scientific

039200-500MG
101847-018EA 193.85 USD
101847-018
[(2-Oxo-2-piperidin-1-ylethyl)(phenyl)amino]acetic acid
[(2-Oxo-2-piperidin-1-ylethyl)(phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₃
MDL Number: MFCD00563289
CAS Number: 858967-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR