[(2-Oxo-2-pyrrolidin-1-ylethyl)(phenyl)amino]acetic acid

Supplier: Matrix Scientific

039198-500MG
101847-014EA 144.9 USD
101847-014
[(2-Oxo-2-pyrrolidin-1-ylethyl)(phenyl)amino]acetic acid
[(2-Oxo-2-pyrrolidin-1-ylethyl)(phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MDL Number: MFCD12027702
CAS Number: 1142211-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR