[[2-(4-Benzylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039197-500MG
101847-012EA 193.85 USD
101847-012
[[2-(4-Benzylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid
[[2-(4-Benzylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₂₁H₂₅N₃O₃
MDL Number: MFCD12027701
CAS Number: 1142211-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR