[[2-(4-Acetylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039192-500MG
101847-002EA 193.85 USD
101847-002
[[2-(4-Acetylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid
[[2-(4-Acetylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₆H₂₁N₃O₄
MDL Number: MFCD12027696
CAS Number: 1142211-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR