[[2-Oxo-2-(4-pyridin-2-ylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039189-500MG
101846-996EA 193.85 USD
101846-996
[[2-Oxo-2-(4-pyridin-2-ylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid
[[2-Oxo-2-(4-pyridin-2-ylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₉H₂₂N₄O₃
MDL Number: MFCD12027693
CAS Number: 1142211-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR