[[2-Oxo-2-(4-pyrimidin-2-ylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039188-500MG
101846-994EA 144.9 USD
101846-994
[[2-Oxo-2-(4-pyrimidin-2-ylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid
[[2-Oxo-2-(4-pyrimidin-2-ylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₈H₂₁N₅O₃
MDL Number: MFCD12027692
CAS Number: 1142211-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR