[[2-Oxo-2-(4-phenylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039185-500MG
101846-988EA 144.9 USD
101846-988
[[2-Oxo-2-(4-phenylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid
[[2-Oxo-2-(4-phenylpiperazin-1-yl)ethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₂₀H₂₃N₃O₃
MDL Number: MFCD12027689
CAS Number: 1142206-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR