[[2-(4-Methylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid
Supplier: Matrix Scientific
039182-500MG
101846-982EA
193.85
USD
101846-982
[[2-(4-Methylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid
[[2-(4-Methylpiperazin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid
Hazard: Irritant
Formula:
C₁₅H₂₁N₃O₃ |
MDL Number:
MFCD12027686 CAS Number: 1142205-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...