[[2-(Diethylamino)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039179-500MG
101846-978EA 144.9 USD
101846-978
[[2-(Diethylamino)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid
[[2-(Diethylamino)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₄
MDL Number: MFCD12027683
CAS Number: 1142205-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR