[[2-(2,6-Dimethylmorpholin-4-yl)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039174-500MG
101846-968EA 193.85 USD
101846-968
[[2-(2,6-Dimethylmorpholin-4-yl)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid
[[2-(2,6-Dimethylmorpholin-4-yl)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₅
MDL Number: MFCD12027678
CAS Number: 1142205-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR