[(4-Methoxyphenyl)(2-oxo-2-thiomorpholin-4-ylethyl)amino]acetic acid

Supplier: Matrix Scientific

039165-500MG
101846-950EA 144.9 USD
101846-950
[(4-Methoxyphenyl)(2-oxo-2-thiomorpholin-4-ylethyl)amino]acetic acid
[(4-Methoxyphenyl)(2-oxo-2-thiomorpholin-4-ylethyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₄S
MDL Number: MFCD12027669
CAS Number: 1142205-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR