[[2-(4-Ethylpiperazin-1-yl)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039147-500MG
101846-916EA 144.9 USD
101846-916
[[2-(4-Ethylpiperazin-1-yl)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid
[[2-(4-Ethylpiperazin-1-yl)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₄
MDL Number: MFCD12027651
CAS Number: 1142205-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR