[[2-(Cycloheptylamino)-2-oxoethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039140-500MG
101846-902EA 193.85 USD
101846-902
[[2-(Cycloheptylamino)-2-oxoethyl](phenyl)amino]acetic acid
[[2-(Cycloheptylamino)-2-oxoethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₃
MDL Number: MFCD12027644
CAS Number: 1142205-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR