[[2-(Cyclopropylamino)-2-oxoethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039128-500MG
101846-878EA 193.85 USD
101846-878
[[2-(Cyclopropylamino)-2-oxoethyl](phenyl)amino]acetic acid
[[2-(Cyclopropylamino)-2-oxoethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD12027632
CAS Number: 1142205-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR