[[2-(Cycloheptylamino)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039098-500MG
101846-818EA 193.85 USD
101846-818
[[2-(Cycloheptylamino)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid
[[2-(Cycloheptylamino)-2-oxoethyl](4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₄
MDL Number: MFCD12027602
CAS Number: 1142204-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR