3-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzoic acid

Supplier: Matrix Scientific

038068-500MG
101844-748EA 290.77 USD
101844-748
3-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzoic acid
3-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₇NO₄
MDL Number: MFCD12027271
CAS Number: 876294-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR