2-(4-Ethylphenyl)-7-methylimidazo[1,2-a]pyridine-3-carbaldehyde

Supplier: Matrix Scientific

038064-500MG
101844-740EA 387.69 USD
101844-740
2-(4-Ethylphenyl)-7-methylimidazo[1,2-a]pyridine-3-carbaldehyde
2-(4-Ethylphenyl)-7-methylimidazo[1,2-a]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₆N₂O
MDL Number: MFCD05177734
CAS Number: 899360-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR