6-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carbaldehyde

Supplier: Matrix Scientific

038058-500MG
101844-728EA 289.8 USD
101844-728
6-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carbaldehyde
6-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₁₂N₂O
MDL Number: MFCD04117107
CAS Number: 727975-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR