4-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzoic acid

Supplier: Matrix Scientific

038055-500MG
101844-720EA 217.35 USD
101844-720
4-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzoic acid
4-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₇NO₄
MDL Number: MFCD12027270
CAS Number: 500728-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR