2-Phenylimidazo[1,2-a]pyridine-3-carbaldehyde

Supplier: Matrix Scientific

038053-500MG
101844-716EA 289.8 USD
101844-716
2-Phenylimidazo[1,2-a]pyridine-3-carbaldehyde
2-Phenylimidazo[1,2-a]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀N₂O
MDL Number: MFCD00964691
CAS Number: 3672-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR