2,8-Dimethyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole

Supplier: Matrix Scientific

038047-500MG
101844-704EA 144.9 USD
101844-704
2,8-Dimethyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole
2,8-Dimethyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole

Hazard: Irritant

Formula: C₁₃H₁₆N₂
MDL Number: MFCD00452533
CAS Number: 19686-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR