6-(3-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-3-carboxylic acid

Supplier: Matrix Scientific

038045-500MG
101844-700EA 387.69 USD
101844-700
6-(3-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-3-carboxylic acid
6-(3-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₇FN₂O₂S
MDL Number: MFCD11222940
CAS Number: 1119452-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR