1-[2-(1H-Indol-3-yl)ethyl]-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

038032-500MG
101844-676EA 162.15 USD
101844-676
1-[2-(1H-Indol-3-yl)ethyl]-2,5-dimethyl-1H-pyrrole-3-carboxylic acid
1-[2-(1H-Indol-3-yl)ethyl]-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₈N₂O₂
MDL Number: MFCD07800638
CAS Number: 893772-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR