Boc-S-4-methylbenzyl-D-cysteine
Supplier: Matrix Scientific
Synonyms:
Boc-D-Cys(pMeBzl)-OH
, Boc-D-Cys(Mbzl)-OH
038022-500MG
101844-656EA
193.85
USD
101844-656
Boc-S-4-methylbenzyl-D-cysteine
Boc-S-4-methylbenzyl-D-cysteine
CAS: 61925-78-8
MDL Number: MFCD01823326
Molecular Formula: C16H23NO4S
Molecular Weight: 325,43
Hazard: Irritant
Formula:
C₁₆H₂₃NO₄S MW: 325.423 g/mol |
MDL Number:
MFCD00076924 CAS Number: 61925-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...