3-(1-Adamantyloxy)propan-1-amine
Supplier: Matrix Scientific
038016-500MG
101844-644EA
336.92
USD
101844-644
3-(1-Adamantyloxy)propan-1-amine
3-(1-Adamantyloxy)propan-1-amine
Hazard: Irritant
Formula:
C₁₃H₂₃NO |
MDL Number:
MFCD02266674 CAS Number: 21624-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...