3-[3-(Ethoxycarbonyl)-5-(4-fluorophenyl)-2-methyl-1H-pyrrol-1-yl]propanoic acid

Supplier: Matrix Scientific

038014-500MG
101844-640EA 290.77 USD
101844-640
3-[3-(Ethoxycarbonyl)-5-(4-fluorophenyl)-2-methyl-1H-pyrrol-1-yl]propanoic acid
3-[3-(Ethoxycarbonyl)-5-(4-fluorophenyl)-2-methyl-1H-pyrrol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₇H₁₈FNO₄
Melting Pt: 107…108 °C
MDL Number: MFCD12027260
CAS Number: 879329-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR