2-[5-(Bromomethyl)-2-ethoxyphenyl]-2H-1,2,3-benzotriazole

Supplier: Matrix Scientific

038010-500MG
101844-632EA 290.77 USD
101844-632
2-[5-(Bromomethyl)-2-ethoxyphenyl]-2H-1,2,3-benzotriazole
2-[5-(Bromomethyl)-2-ethoxyphenyl]-2H-1,2,3-benzotriazole

Hazard: Irritant

Formula: C₁₅H₁₄BrN₃O
MDL Number: MFCD12027258
CAS Number: 1119451-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR