5-Phenyl-6H-1,3,4-thiadiazin-2-amine
Supplier: Matrix Scientific
037998-500MG
101844-608EA
217.35
USD
101844-608
5-Phenyl-6H-1,3,4-thiadiazin-2-amine
5-Phenyl-6H-1,3,4-thiadiazin-2-amine
Hazard: Irritant
Formula:
C₉H₉N₃S |
MDL Number:
MFCD00524799 CAS Number: 55185-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...