4-Nitro-1,2,5-oxadiazol-3-amine

Supplier: Matrix Scientific

037987-500MG
101844-588EA 290.77 USD
101844-588
4-Nitro-1,2,5-oxadiazol-3-amine
4-Nitro-1,2,5-oxadiazol-3-amine

Hazard: Irritant

Formula: C₂H₂N₄O₃
MDL Number: MFCD00186405
CAS Number: 66328-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR