3-[3-(Ethoxycarbonyl)-2-methyl-5-(4-methylphenyl)-1H-pyrrol-1-yl]propanoic acid

Supplier: Matrix Scientific

037976-500MG
101844-566EA 290.77 USD
101844-566
3-[3-(Ethoxycarbonyl)-2-methyl-5-(4-methylphenyl)-1H-pyrrol-1-yl]propanoic acid
3-[3-(Ethoxycarbonyl)-2-methyl-5-(4-methylphenyl)-1H-pyrrol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₈H₂₁NO₄
MDL Number: MFCD08690580
CAS Number: 396123-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR