3-Amino-3-(2-isopropoxyphenyl)propanoic acid

Supplier: Matrix Scientific

037950-500MG
101844-514EA 144.9 USD
101844-514
3-Amino-3-(2-isopropoxyphenyl)propanoic acid
3-Amino-3-(2-isopropoxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₇NO₃
MDL Number: MFCD12027249
CAS Number: 890045-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR