(3,5,6-Trifluoropyridin-2-yl)acetic acid

Supplier: Matrix Scientific

037948-500MG
101844-510EA 217.35 USD
101844-510
(3,5,6-Trifluoropyridin-2-yl)acetic acid
(3,5,6-Trifluoropyridin-2-yl)acetic acid

Hazard: Irritant

Formula: C₇H₄F₃NO₂
MDL Number: MFCD03001145
CAS Number: 1119450-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR