3-Amino-3-(4-isobutoxyphenyl)propanoic acid

Supplier: Matrix Scientific

037938-500MG
101844-490EA 144.9 USD
101844-490
3-Amino-3-(4-isobutoxyphenyl)propanoic acid
3-Amino-3-(4-isobutoxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₃
MDL Number: MFCD00297955
CAS Number: 383373-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR