2-(Methylsulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Supplier: Matrix Scientific

037936-500MG
101844-486EA 289.8 USD
101844-486
2-(Methylsulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
2-(Methylsulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄S
MDL Number: MFCD03021424
CAS Number: 1007984-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR