1-Oxo-4-phenyl-1H-isochromene-3-carboxylic acid

Supplier: Matrix Scientific

037932-500MG
101844-478EA 251.85 USD
101844-478
1-Oxo-4-phenyl-1H-isochromene-3-carboxylic acid
1-Oxo-4-phenyl-1H-isochromene-3-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₀O₄
MDL Number: MFCD02853676
CAS Number: 37617-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR