(2E)-3-[4-(2-Amino-2-oxoethoxy)-3-methoxyphenyl]acrylic acid

Supplier: Matrix Scientific

037930-500MG
101844-474EA 144.9 USD
101844-474
(2E)-3-[4-(2-Amino-2-oxoethoxy)-3-methoxyphenyl]acrylic acid
(2E)-3-[4-(2-Amino-2-oxoethoxy)-3-methoxyphenyl]acrylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₅
MDL Number: MFCD02256283
CAS Number: 696645-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR