3-Amino-3-(4-chloro-3-nitrophenyl)propanoic acid

Supplier: Matrix Scientific

037928-500MG
101844-470EA 193.85 USD
101844-470
3-Amino-3-(4-chloro-3-nitrophenyl)propanoic acid
3-Amino-3-(4-chloro-3-nitrophenyl)propanoic acid

Hazard: Irritant

Formula: C₉H₉ClN₂O₄
MDL Number: MFCD03002540
CAS Number: 682804-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR