3-Ethoxy-4-(3-methylbutoxy)benzaldehyde

Supplier: Matrix Scientific

037926-500MG
101844-466EA 109.25 USD
101844-466
3-Ethoxy-4-(3-methylbutoxy)benzaldehyde
3-Ethoxy-4-(3-methylbutoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₄H₂₀O₃
MDL Number: MFCD10007539
CAS Number: 85259-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR