2-(Chloromethyl)-4H-pyrido[1,2-a]pyrimidin-4-one

Supplier: Matrix Scientific

037922-500MG
101844-458EA 95.38 USD
101844-458
2-(Chloromethyl)-4H-pyrido[1,2-a]pyrimidin-4-one
2-(Chloromethyl)-4H-pyrido[1,2-a]pyrimidin-4-one

Hazard: Irritant

Formula: C₉H₇ClN₂O
MDL Number: MFCD03001117
CAS Number: 16867-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR