1-(3-Nitrophenyl)-1H-pyrrole-2-carbaldehyde

Supplier: Matrix Scientific

037920-500MG
101844-454EA 193.85 USD
101844-454
1-(3-Nitrophenyl)-1H-pyrrole-2-carbaldehyde
1-(3-Nitrophenyl)-1H-pyrrole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₈N₂O₃
MDL Number: MFCD00453253
CAS Number: 85841-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR