6-Bromo-2-(2-furyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

037918-500MG
101844-450EA 165.6 USD
101844-450
6-Bromo-2-(2-furyl)quinoline-4-carboxylic acid
6-Bromo-2-(2-furyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₈BrNO₃
MDL Number: MFCD01141800
CAS Number: 296244-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR