3-(1,3-Dioxo-1,3,3a,4,7,7a-hexahydro-2H-isoindol-2-yl)benzoic acid

Supplier: Matrix Scientific

037916-500MG
101844-446EA 146.15 USD
101844-446
3-(1,3-Dioxo-1,3,3a,4,7,7a-hexahydro-2H-isoindol-2-yl)benzoic acid
3-(1,3-Dioxo-1,3,3a,4,7,7a-hexahydro-2H-isoindol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃NO₄
MDL Number: MFCD00523810
CAS Number: 89446-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR