[(7-Chloro-4-nitro-2,1,3-benzoxadiazol-5-yl)thio]acetic acid

Supplier: Matrix Scientific

037914-500MG
101844-442EA 336.92 USD
101844-442
[(7-Chloro-4-nitro-2,1,3-benzoxadiazol-5-yl)thio]acetic acid
[(7-Chloro-4-nitro-2,1,3-benzoxadiazol-5-yl)thio]acetic acid

Hazard: Irritant

Formula: C₈H₄ClN₃O₅S
MDL Number: MFCD02656525
CAS Number: 442531-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR