5-Nitro-1-phenyl-1H-indazol-3-amine
Supplier: Matrix Scientific
037912-500MG
101844-438EA
387.69
USD
101844-438
5-Nitro-1-phenyl-1H-indazol-3-amine
5-Nitro-1-phenyl-1H-indazol-3-amine
Hazard: Irritant
Formula:
C₁₃H₁₀N₄O₂ |
MDL Number:
MFCD02111866 CAS Number: 309944-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...