2-(3,5-Dimethylbenzoyl)benzoic acid
Supplier: Matrix Scientific
037908-500MG
101844-430EA
290.77
USD
101844-430
2-(3,5-Dimethylbenzoyl)benzoic acid
2-(3,5-Dimethylbenzoyl)benzoic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₃ |
MDL Number:
MFCD00429472 CAS Number: 149817-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...