4-Anilino-1-benzylpiperidine-4-carboxamide

Supplier: Matrix Scientific

037890-500MG
101844-390EA 461.54 USD
101844-390
4-Anilino-1-benzylpiperidine-4-carboxamide
4-Anilino-1-benzylpiperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₉H₂₃N₃O
MDL Number: MFCD00474726
CAS Number: 1096-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR