Isatin-3-oxime

Supplier: Matrix Scientific

037882-500MG
101844-374EA 110.77 USD
101844-374
Isatin-3-oxime
Isatin-3-oxime

Hazard: Irritant

Formula: C₈H₆N₂O₂
Melting Pt: 217 °C
MDL Number: MFCD00014568
CAS Number: 607-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR