6-Chloro-N,N-dimethyl-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

037880-500MG
101844-370EA 217.35 USD
101844-370
6-Chloro-N,N-dimethyl-1,3,5-triazine-2,4-diamine
6-Chloro-N,N-dimethyl-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₅H₈ClN₅
MDL Number: MFCD00460497
CAS Number: 32998-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR