5,6-Diphenylfuro[2,3-d]pyrimidin-4-amine

Supplier: Matrix Scientific

037876-500MG
101844-362EA 217.35 USD
101844-362
5,6-Diphenylfuro[2,3-d]pyrimidin-4-amine
5,6-Diphenylfuro[2,3-d]pyrimidin-4-amine

Hazard: Irritant

Formula: C₁₈H₁₃N₃O
MDL Number: MFCD00407207
CAS Number: 5207-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR