4-[(Diethylamino)sulfonyl]benzoic acid

Supplier: Matrix Scientific

037874-500MG
101844-358EA 217.35 USD
101844-358
4-[(Diethylamino)sulfonyl]benzoic acid
4-[(Diethylamino)sulfonyl]benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₅NO₄S
MDL Number: MFCD00020373
CAS Number: 1213-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR